5-Methoxy-2-methylindole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1076-74-0
:
Formula:C₁₀H₁₁NO
MDL Number:MFCD00005620
Boiling Pt:145 °C (1.5 mmHg)
Melting Pt:89 °C
:M-Clarity™ quality level = MQ100
:161.2 g/mol
EINECS:214-066-5
Frequently Bought Together