1,3-Dibutyl-2-thiourea, Sigma-Aldrich®
Catalog # SIALD49598-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:188.34 g/mol
Boiling Pt:252 °C (1013 hPa)
Melting Pt:62…65 °C
:M-Clarity™ quality level = MQ100
EINECS:203-674-6
CAS Number:109-46-6
Formula:H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
MDL Number:MFCD00004926