4-(Aminomethyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3731-53-1
:
Formula:C₆H₈N₂
MDL Number:MFCD00006449
:108.14 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:–8 °C
Density:1.0688 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:223-092-6
Frequently Bought Together