
Sodium deoxycholate
Catalog # SIAL96949-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:414.56 g/mol
Melting Pt:357…365 °C
:M-Clarity™ quality level = MQ200
EINECS:206-132-7
:GHS07
Storage Temperature:Ambient
CAS Number:302-95-4
Formula:C₂₄H₃₉NaO₄
MDL Number:MFCD00064139