



Palladium(II) nitrate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:32916-07-7
:
Formula:N₂O₆Pd·2H₂O
:266.46 g/mol
EINECS:233-265-8
UN:1477
ADR:5.1,II
:GHS03
:GHS06
:GHS05
:GHS09
Frequently Bought Together