Order Entry
Czech Republic
ContactUsLinkComponent
6-Methyl-2-thiouracil, Sigma-Aldrich®
6-Methyl-2-thiouracil, Sigma-Aldrich®
Catalog # SIAL69400-100G
Supplier:  Merck
CAS Number:  
6-Methyl-2-thiouracil, Sigma-Aldrich®
Catalog # SIAL69400-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :142.18 g/mol
Merck Index:12,06210
Boiling Pt:351 °C (1013 hPa)
Density:1.519 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
EINECS:200-252-3
CAS Number:56-04-2
 :MFCD00006040
Formula:C₅H₆N₂OS