4-Iodo-2-nitroaniline, Sigma-Aldrich®
Catalog # SIAL654108-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.02 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:20691-72-9
Formula:C₆H₅IN₂O₂
MDL Number:MFCD06808526