Order Entry
Czech Republic
ContactUsLinkComponent
2-Propanol-[D1], Sigma-Aldrich®
Flammable
Irritant
2-Propanol-[D1], Sigma-Aldrich®
Catalog # SIAL615080-5G
Supplier:  Merck
CAS Number:  
Flammable
Irritant
2-Propanol-[D1], Sigma-Aldrich®
Catalog # SIAL615080-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :61.09 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89.5 °C
Density:0.798 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:223-616-3
UN:1219
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
CAS Number:3979-51-9
Formula:C₃H₇DO