Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, Isoprene
Supelco® Organic Reference Standard, Isoprene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Isoprene
Supplier:  Merck

Specifications

  • CAS No.:
    78-79-5
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H224 - H341 - H350 - H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008600
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Methyl-1,3-butadiene
  • Melting point:
    –146 °C (lit.)
  • Formula:
    CH₂=CHC(CH₃)=CH₂
  • Molecular weight:
    68,12
  • Density:
    0,681 g/ml at 20 °C; 0,681 g/ml at 25 °C (lit.)
  • Flash point:
    –54 °C
  • Boiling point:
    34 °C (lit.)

Specifications

Frequently Bought Together