Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2,3,3-Tetrafluoro-1,4-butanediol, Sigma-Aldrich®
Katalogové číslo SIAL567191-5G
Supplier:  Merck
CAS Number:  
2,2,3,3-Tetrafluoro-1,4-butanediol, Sigma-Aldrich®
Katalogové číslo SIAL567191-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :162.08 g/mol
Boiling Pt:110…112 °C (13 mmHg)
Melting Pt:85 °C
 :M-Clarity™ quality level = MQ100
CAS Number:425-61-6
Formula:C₄H₆F₄O₂
MDL Number:MFCD00042375