3-(Diethylamino)propyl isothiocyanate, Sigma-Aldrich®
SIAL560510-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:172.29 g/mol
:88…92 °C (0.1 mmHg)
:0.9535 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:2626-52-0
:C₈H₁₆N₂S
:MFCD00041133