1-Butyl-3-methylimidazolium methyl sulphate, Sigma-Aldrich®
Catalog # SIAL53177-100G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:250.32 g/mol
Density:1.21 g/cm³ (20 °C)
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
CAS Number:401788-98-5
Formula:C₉H₁₈N₂O₄S
MDL Number:MFCD03095437