
2-(Trifluoromethoxy)benzaldehyde, Sigma-Aldrich®
Catalog # SIAL529168-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:190.12 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:94651-33-9
Formula:C₈H₅F₃O₂
MDL Number:MFCD00042405