Order Entry
Czech Republic
ContactUsLinkComponent
(Methyl)triphenylphosphonium chloride, Sigma-Aldrich®
Corrosive
Irritant
Environment
(Methyl)triphenylphosphonium chloride, Sigma-Aldrich®
Catalog # SIAL468002-5G
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Environment
(Methyl)triphenylphosphonium chloride, Sigma-Aldrich®
Catalog # SIAL468002-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :312.78 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:418-400-2
 :GHS05
 :GHS07
 :GHS09
CAS Number:1031-15-8
Formula:C₁₉H₁₈ClP
MDL Number:MFCD00797851