Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Dimethyl-1,1,1-trimethylsilylamine, Supelco®
N,N-Dimethyl-1,1,1-trimethylsilylamine, Supelco®
Catalog # 41716-5ML
Supplier:  Merck
CAS Number:  
undefined
N,N-Dimethyl-1,1,1-trimethylsilylamine, Supelco®
Catalog # 41716-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :117.27 g/mol
Boiling Pt:84 °C (1013 hPa)
Density:0.7466 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:218-222-3
CAS Number:2083-91-2
Formula:C₅H₁₅NSi
MDL Number:MFCD00008297