


Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7226-23-5
:
Formula:C₆H₁₂N₂O
MDL Number:MFCD00006550
:128.17 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:121 °C
EINECS:230-625-6
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together