Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
di-Ammonium oxalate monohydrate, Sigma-Aldrich®
Catalog # SIAL379743-25G
Supplier:  Merck
CAS Number:  
Irritant
di-Ammonium oxalate monohydrate, Sigma-Aldrich®
Catalog # SIAL379743-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-202-3
UN:3288
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:6009-70-7
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694