(2-Methoxyethoxy)methyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3970-21-6
:
Formula:ClCH₂OCH₂CH₂OCH₃
MDL Number:MFCD00000888
:124.57 g/mol
Boiling Pt:50 °C (1013 hPa)
Density:1.094 g/cm³ (20 °C)
EINECS:223-589-8
UN:1992
ADR:3,III
:M-Clarity™ quality level = MQ200
Frequently Bought Together