Tripropyl orthoformate, Sigma-Aldrich®
Catalog # SIAL333344-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:190.28 g/mol
:M-Clarity™ quality level = MQ100
EINECS:210-704-1
CAS Number:621-76-1
Formula:C₁₀H₂₂O₃
MDL Number:MFCD00015214