Sodium deoxycholate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:302-95-4
:
Formula:C₂₄H₃₉NaO₄
MDL Number:MFCD00064139
:414.56 g/mol
Melting Pt:357…365 °C
EINECS:206-132-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together