2,6-Dimethyl-5-heptenal, Sigma-Aldrich®
Katalogové číslo SIAL307963-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:140.23 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:203-427-2
UN:1989
CAS Number:106-72-9
Formula:C₉H₁₆O
MDL Number:MFCD00006981