Tris(pentafluorophenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL290572-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:532.15 g/mol
:M-Clarity™ quality level = MQ200
EINECS:215-021-2
CAS Number:1259-35-4
Formula:C₁₈F₁₅P
MDL Number:MFCD00079654