
Dysprosium trichloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL289272-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:376.95
Melting Pt:163 °C
:M-Clarity™ quality level = MQ100
EINECS:233-039-9
:GHS07
CAS Number:15059-52-6
Formula:Cl₃Dy·6H₂O
MDL Number:MFCD00010984