


Disodium hexachloroplatinate hexahydrate, Sigma-Aldrich®
Catalog # SIAL288152-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Merck Index:12,08768
Density:2.5 g/cm³ (20 °C)
EINECS:240-983-5
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:561.87 g/mol
UN:3288
:GHS06
:GHS05
:GHS08
CAS Number:19583-77-8
Formula:Cl₆Na₂Pt·6H₂O
MDL Number:MFCD00149172