tert-Butyl ethyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:637-92-3
:
Formula:C₆H₁₄O
MDL Number:MFCD00009225
:102.18 g/mol
Merck Index:13,03809
Boiling Pt:71 °C (1013 hPa)
Melting Pt:–97 °C
Density:0.752 g/cm³ (20 °C)
UN:1179
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:211-309-7
Frequently Bought Together