Tetrakis(triphenylphosphine)nickel(0), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15133-82-1
:
Formula:C₇₂H₆₀NiP₄
MDL Number:MFCD00010011
:1107.85 g/mol
ADR:4.1,II
:M-Clarity™ quality level = MQ100
UN:1325
Frequently Bought Together