trans,trans,trans-1,5,9-Cyclododecatriene, Sigma-Aldrich®
SIAL234273-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:162.27 g/mol
:237…238 °C (1013 hPa)
:35…37 °C
:6.1,III
:M-Clarity™ quality level = MQ100
:211-626-0
:2518
:676-22-2
:C₁₂H₁₈
:MFCD00003720