Order Entry
Czech Republic
ContactUsLinkComponent
(3-Methoxyphenyl)acetonitrile, Sigma-Aldrich®
(3-Methoxyphenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIAL209392-5G
Supplier:  Merck
CAS Number:  
(3-Methoxyphenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIAL209392-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:164 °C (1013 hPa)
Density:1.05 g/cm³ (20 °C)
EINECS:243-428-5
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :147.18 g/mol
UN:3276
CAS Number:19924-43-7
Formula:C₉H₉NO
MDL Number:MFCD00001910