Disodium hexachloroiridate hexahydrate, Sigma-Aldrich®
Catalog # SIAL209007-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:559.01 g/mol
Melting Pt:600 °C
:M-Clarity™ quality level = MQ100
EINECS:241-011-2
CAS Number:19567-78-3
Formula:Cl₆IrNa₂·6H₂O
MDL Number:MFCD00149169