Order Entry
Czech Republic
ContactUsLinkComponent
1,1,4,4-Tetraphenyl-1,3-butadiene, Sigma-Aldrich®
Irritant
1,1,4,4-Tetraphenyl-1,3-butadiene, Sigma-Aldrich®
Catalog # SIAL185213-5G
Supplier:  Merck
CAS Number:  
Irritant
1,1,4,4-Tetraphenyl-1,3-butadiene, Sigma-Aldrich®
Catalog # SIAL185213-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :358.48 g/mol
Boiling Pt:556 °C (1013 hPa)
Melting Pt:207…209 °C
 :M-Clarity™ quality level = MQ100
EINECS:215-914-7
 :GHS07
CAS Number:1450-63-1
Formula:(C₆H₅)₂CCHCHC(C₆H₅)₂
MDL Number:MFCD00004766