Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2-Hydroxyethyl)phthalimide
N-(2-Hydroxyethyl)phthalimide, Sigma-Aldrich®
N-(2-Hydroxyethyl)phthalimide, Sigma-Aldrich®
SIAL138339-100G
:
Merck
:
N-(2-Hydroxyethyl)phthalimide, Sigma-Aldrich®
SIAL138339-100G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
191.19 g/mol
:
223-434-4
:
3891-07-4
:
C₁₀H₉NO₃
:
MFCD00005903