3-Methyl-1,2-butadiene, Sigma-Aldrich®
Catalog # SIAL110930-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:68.12 g/mol
Boiling Pt:40 °C (1013 hPa)
Melting Pt:–113.6 °C
Density:0.6831 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:209-926-1
CAS Number:598-25-4
Formula:C₅H₈
MDL Number:MFCD00008896