
2-Bromethylamine hydrobromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2576-47-8
:
Formula:BrCH₂CH₂NH₃Br
MDL Number:MFCD00012886
:204.89 g/mol
Melting Pt:170…173 °C
:M-Clarity™ quality level = MQ200
EINECS:219-924-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together