
6-Propyl-2-thiouracil
Katalogové číslo PROOICRS0403
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:170.24 g/mol
Merck Index:12,08054
Boiling Pt:448 °C (1013 hPa)
Melting Pt:215…218 °C
EINECS:200-103-2
:GHS07
CAS Number:51-52-5
Formula:C₇H₁₀N₂OS
MDL Number:MFCD00006041