Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,6-Dichlorophenyl)-2-thiourea
Toxic
1-(2,6-Dichlorophenyl)-2-thiourea
Catalog # PROOMM0983.02
CAS Number:  
Toxic
1-(2,6-Dichlorophenyl)-2-thiourea
Catalog # PROOMM0983.02
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :221.11 g/mol
Boiling Pt:326 °C (1013 hPa)
Melting Pt:160…162 °C
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:6590-91-6
Formula:(C₆H₃Cl₂)NC(S)NH₂
MDL Number:MFCD00041156