4'-Isopropylacetophenone
Catalog # PROOMM0002.57
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Density:0.97 g/cm³ (20 °C)
:162.23
EINECS:211-433-1
CAS Number:645-13-6
Formula:C₁₁H₁₄O
MDL Number:MFCD00048297