Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Octadecyltrichlorosilane, Sigma-Aldrich®
  8.22170.0100
 :  Merck
 :  
 
 
Octadecyltrichlorosilane, Sigma-Aldrich®
  8.22170.0100
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

 

 

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    387.94 g/mol
  • Boiling Point:
    223 °C (13 hPa)
  • Flash Point:
    89 °C
  • Density:
    1228 g/mol
  • pH:
    (H2O, 20 °C) not applicable
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

 

 :
 :387.94 g/mol
 :361 °C (1013 hPa)
 :22 °C
 :0.98 g/cm³ (20 °C)
 :89 °C
 :Ambient
 :203-930-7
 :1800
 :8,II
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS07
 :112-04-9
 :C₁₈H₃₇Cl₃Si
 :MFCD00000484