
1,3-Dibutyl-2-thiourea, Sigma-Aldrich®
8.20423.0250
: Merck
:
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Solubility:(20 °C) practically insoluble
- Melting Point:62 - 65 °C
- Molar Mass:188.34 g/mol
- Bulk Density:500 kg/m3
- Vapor Pressure:0.0032 hPa (25 °C)
- pH:6 (0.2 g/l, H2O, 20 °C)
- Hill Formula:C9H20N2S
- Chemical Formula:(CH3CH2CH2CH2NH)2CS
:
:188.34 g/mol
:252 °C (1013 hPa)
:62…65 °C
:M-Clarity™ quality level = MQ200
:Ambient
:203-674-6
:GHS07
:109-46-6
:H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
:MFCD00004926