


Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7226-23-5
:
Formula:C₆H₁₂N₂O
MDL Number:MFCD00006550
:128.17 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.06 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:121 °C
EINECS:230-625-6
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20°C) Soluble
- Melting Point:-24 °C
- Boiling Point:247 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (57 °C)
- Flash Point:121 °C
- Explosion Limit:1.6 - 9.3 %(V)
- Density:1.064 g/cm3 (20 °C)
- pH:11 (100 g/l, H2O, 20 °C)
- Ignition Point:255°C
- Synonyms:Dimethylpropyleneurea, DMPU
- Hill Formula:C6H12N2O
Specification Test Results
Frequently Bought Together