4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221) ≥98% for synthesis, Sigma-Aldrich®
8.10646.0001
: Merck
:
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
- Purity (GC):≥ 98
- Identity (IR-spectrum):passes test
- Density (d 20 °C/ 4 °C):1.107 - 1.114
- Water (according to Karl Fischer):≤ 0.5
:
:332.44 g/mol
:1.11 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:250-592-1
:31364-42-8
:C₁₆H₃₂N₂O₅
:MFCD00005108