Order Entry
Czech Republic
ContactUsLinkComponent
2,2'-Methyliminodiethanol for synthesis
Irritant
2,2'-Methyliminodiethanol for synthesis
Supplier:  Merck
Certificates
Irritant
2,2'-Methyliminodiethanol for synthesis
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -21 °C
  • Molar Mass:
    119.16 g/mol
  • Boiling Point:
    243 °C (1013 hPa)
  • Vapor Pressure:
    0.026 hPa (40 °C)
  • Flash Point:
    137 °C
  • Refractive Index:
    1.4685 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 - 8.4 %(V)
  • Density:
    1.04 g/cm3 (20 °C)
  • pH:
    11.5 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    265 °C

Specification Test Results

Related Information

Frequently Bought Together