


1,3-Diphenyl-2-thiourea, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:102-08-9
:
Formula:C₁₃H₁₂N₂S
MDL Number:MFCD00004921
:228.32 g/mol
Merck Index:13,03366
Boiling Pt:365 °C (1013 hPa)
Melting Pt:150…153 °C
Density:1.3205 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:203-004-2
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:150 - 153 °C
- Molar Mass:228.31 g/mol
- Bulk Density:400 kg/m3
- Density:1.30 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together