Order Entry
Czech Republic
ContactUsLinkComponent
Trimethyl orthoformate, Sigma-Aldrich®
Flammable
Irritant
Trimethyl orthoformate, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Trimethyl orthoformate, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    10 g/l (20 °C) Hydrolysis
  • Melting Point:
    -53 °C
  • Molar Mass:
    106.12 g/mol
  • Boiling Point:
    102 °C (1013 hPa)
  • Vapor Pressure:
    10 hPa (7 °C)
  • Flash Point:
    13 °C
  • Refractive Index:
    1.379 (20 °C, 589 nm)
  • Explosion Limit:
    1.4 - 44.6 %(V)
  • Density:
    0.97 g/cm3 (20 °C)
  • Ignition Point:
    255 °C

Specification Test Results

Related Information

Frequently Bought Together