Uhličitan sodný 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2,53 g/cm³
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Specification Test Results
- Meets N.F. Requirements:Passes Test
- Meets J.P. Chemical Specifications:Passes Test
- Meets B.P. Chemical Specifications:≤0.5%
- Meets E.P. Chemical Specifications:99.5 - 100.5%
- GMP Manufactured Product:99.5 - 100.5%
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Water (H₂O):≤ 0.5 %
- NF - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Assay (Na₂CO₃) (dried basis):99.5 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Identification D:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Alkali Hydroxides and Bicarbonates:Passes Test
- EP/BP - Chloride (Cl):≤ 125 ppm
- EP/BP - Sulfate (SO₄):≤ 250 ppm
- EP/BP - Iron (Fe):≤ 50 ppm
- EP/BP - Loss on Drying:≤ 1.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.071 %
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Arsenic (As):≤ 3.1 ppm
- JP - Loss on Drying:≤ 0.5 %
- JP - Assay (Na₂CO₃) (dried basis):99.0 - 101.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR3606-07 spec_BAKR3606-09 spec_BAKR3606-R
Product specification
Frequently Bought Together