Order Entry
Czech Republic
ContactUsLinkComponent
EDTA dipotassium salt dihydrate, Ultrapure
EDTA dipotassium salt dihydrate, Ultrapure
Catalog # 0720-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA dipotassium salt dihydrate, Ultrapure
Catalog # 0720-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Chloride:
    ≤0.005%
  • Heavy Metals:
    ≤0.001%
  • Iron:
    ≤0.001%
  • pH (5%, Water) @25 °C:
    4.0 – 5.0
  • Purity:
    ≥99.0%
  • Appearance:
    White granular powder

Specification Test Results

About this item

 :
 :404.46 g/mol
Merck Index:13,03546
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-895-0
CAS Number:25102-12-9
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MDL Number:MFCD00150036