
1-Ethyl-3-methylimidazolium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65039-08-9
:
Formula:C₆H₁₁BrN₂
MDL Number:MFCD03427610
Melting Pt:70…73 °C
:191.07 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together