
1,3-Difluoro-5-iodobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2265-91-0
:
Formula:C₆H₃F₂I
MDL Number:MFCD03094155
:239.99 g/mol
Boiling Pt:173…174 °C
Flash Pt:82 °C (179 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together