Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
2-Chloro-3,6-difluorobenzylamine 97%
Catalog # L19140.03
CAS Number:  
Corrosive
2-Chloro-3,6-difluorobenzylamine 97%
Catalog # L19140.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:261762-45-2
 :
Formula:C₇H₆ClF₂N
MDL Number:MFCD01631449
 :177.58 g/mol
Boiling Pt:207 °C (1013 hPa)
Melting Pt:73 °C
Storage Temperature:Ambient
ADR:8,II
 :GHS05