cis-6-Nonen-1-ol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35854-86-5
:
Formula:C₉H₁₈O
MDL Number:MFCD00015388
:142.24 g/mol
Boiling Pt:85 °C (5 mmHg)
Density:0.850
Flash Pt:98 °C (208 °F)
EINECS:252-764-1
Frequently Bought Together