

2,2,2-Trichloroethyl chloroformate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17341-93-4
:
Formula:C₃H₂Cl₄O₂
MDL Number:MFCD00000810
:211.86 g/mol
Boiling Pt:171…172 °C
Melting Pt:0…2 °C
Density:1.539
Flash Pt:>100 °C (212 °F)
EINECS:241-363-7
UN:3277
ADR:6.1,II
:GHS06
:GHS05
Related Information
Frequently Bought Together